Acetone 99.5+% for analysis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

176800010 176800025 176805000 176800050 176800250 176800026
ACRO176800010EA 43.5 EUR
ACRO176800010 ACRO176800025 ACRO176805000 ACRO176800050 ACRO176800250 ACRO176800026
Acetone 99.5+% for analysis
Acetone
Formula: CH₃COCH₃
Boiling Pt: 56 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.79 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.5 % (Coulometric)
GC ≥99.50 %
Color scale ≤5 APHA
Residue after evaporation ≤0.001 %
Acidity/alkalinity ≤0.0005 meq/g
Cadmium (Cd) ≤0.05 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Zinc (Zn) ≤0.05 ppm
KMnO4-reducing substance ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR