Acetone 99+%, extra pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

177170010 177170025 177170250 177170050 177170100
ACRO177170010EA 39.9 EUR
ACRO177170010 ACRO177170025 ACRO177170250 ACRO177170050 ACRO177170100
Acetone 99+%, extra pure
Acetone
Formula: CH₃COCH₃
Boiling Pt: 56 °C (760 mmHg)
Melting Pt: –95 °C
Density: 0.79 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.5 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR