Carbon disulphide 99.9%, extra pure low in benzene

Supplier: Thermo Fisher Scientific
Danger

177280010 177280250 177285000 177280025
ACRO177280010EA 127 EUR
ACRO177280010 ACRO177280250 ACRO177285000 ACRO177280025
Carbon disulphide 99.9%, extra pure low in benzene
Carbon disulphide
Formula: CS₂
Boiling Pt: 46.2 °C (1013 hPa)
Melting Pt: –111 °C
Density: 1.262 g/cm³
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Assay ≥99.85 % (By difference)
Residue after evaporation ≤20 ppm
Hydrogen sulfide (H2S) ≤3 ppm
Benzene ≤50 ppm
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR