Oxalic acid, anhydrous 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethanedioic acid

186430010 186432500 186430050
ACRO186430010EA 76.6 EUR
ACRO186430010 ACRO186432500 ACRO186430050
Oxalic acid, anhydrous 98%
Oxalic acid
Formula: C₂H₂O₄
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 189…191 °C
Density: 1.9 g/cm³
Flash Pt: >93.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
ADR: 8,III

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 189 to 191 °C
Titration with NaOH 97.5 to 102.5 %
pH 8.5 to 10.5 (10 % suspension in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR