Basic Blue 24 (New Methylene Blue), pure

Supplier: Thermo Fisher Scientific

Synonyms: New Methylene Blue N, zinc chloride double salt, New Methylene Blue N

192020100
ACRO192020100EA 77.5 EUR
ACRO192020100
Basic Blue 24 (New Methylene Blue), pure
Basic Blue 24 (New Methylene Blue)

Appearance: Dark brown to purple Crystalline powder

Formula: C₁₈H₂₂ClN₃S
Storage Temperature: Ambient
MDL Number: MFCD00078135
CAS Number: 6586-05-6

Order Now

Specification Test Results

Appearance Dark brown to deep purple Crystalline powder
Infrared spectrum Conforms
Dye content ≥90 %
UV Lambda max 242 to 248 nm (c=5 mg/l, water)
Molar extinction coefficient ≥16000 (c=5 mg/l, water) 242 to 248 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR