Nickel(II) chloride hexahydrate 99.999% (trace metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Nickel dichloride hexahydrate

193570050 193570250 193571000
ACRO193570050EA 75.9 EUR
ACRO193570050 ACRO193570250 ACRO193571000
Nickel(II) chloride hexahydrate 99.999% (trace metals basis)
Nickel (II) chloride hexahydrate
Formula: NiCl₂·6H₂O
Melting Pt: 140 °C
Density: 1,84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,06529

Order Now

Specification Test Results

Appearance Green Crystalline powder or crystals and/or chunks
Total trace metal impurities ≤0.001 %
Nickel (Ni) 24 to 26 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR