Copper standard solution, 1000 mg/L Cu in dil. nitric acid standard for AAS
Supplier: Thermo Fisher Scientific
195931000 195935000
ACRO195931000EA 19.4 EUR
ACRO195931000 ACRO195935000
Copper standard solution, 1000 mg/L Cu in dil. nitric acid standard for AAS
Standards Cationic Standards Copper Standards