Silver nitrate 99.85% analysis_for

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

197680050 197680250 197681000 197685000 197680025
ACRO197680050EA 51.5 EUR
ACRO197680050 ACRO197680250 ACRO197681000 ACRO197685000 ACRO197680025
Silver nitrate 99.85% analysis_for
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444 °C (760 mmHg)
Melting Pt: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1 II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Loss on drying ≤0.1 % (1 g, 70°C, 2 h)
Titration with HCl ≥99.80 %
Chloride (Cl) ≤5 ppm
Nitrite (NO2) ≤10 ppm
Cadmium (Cd) ≤1 ppm
Calcium (Ca) ≤10 ppm
Chromium (Cr) ≤1 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Manganese (Mn) ≤1 ppm
Nickel (Ni) ≤1 ppm
Zinc (Zn) ≤1 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR