2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,2-Dimethyl-1,3-dioxane-4,6-dione

198450250 198451000 198455000
ACRO198450250EA 47 EUR
ACRO198450250 ACRO198451000 ACRO198455000
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) 98%
2,2-Dimethyl-1,3-dioxane-4,6-dione (Isopropylidene malonate, Meldrum's acid)
Formula: C₆H₈O₄
Melting Pt: 92…96 °C
Flash Pt: >365 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006638
CAS Number: 2033-24-1
EINECS: 217-992-8
UN: 3077

Order Now

Specification Test Results

Appearance White to beige to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 92 to 96 °C
Water ≤0.1 % (K.F.)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR