Cerium(IV) sulphate tetrahydrate 98+%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cerium disulphate tetrahydrate, Ceric sulphate tetrahydrate

199131000 199135000 199130025
ACRO199131000EA 168 EUR
ACRO199131000 ACRO199135000 ACRO199130025
Cerium(IV) sulphate tetrahydrate 98+%
Cerium (IV) sulphate tetrahydrate
Formula: Ce(SO₄)₂·4H₂O
Melting Pt: 180 - 200 °C
Density: 3,91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149427
CAS Number: 10294-42-5
EINECS: 237-029-5
UN: 3260
ADR: 8,III
Merck Index: 12,02036

Order Now

Specification Test Results

Appearance Bright yellow Powder
Titration Iodometric ≥98.0 %
Chloride (Cl) ≤100 ppm
Copper (Cu) ≤30 ppm
Iron (Fe) ≤150 ppm
Lead (Pb) ≤30 ppm
Nickel (Ni) ≤30 ppm
Heavy metals (as Pb) ≤30 ppm
Insoluble matter ≤0.1 % (in H2SO4)
Subst not pptd by NH4OH ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR