Lead(II) oxide 99.7%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Lead monoxide

199340010 199340025
ACRO199340010EA 67.5 EUR
ACRO199340010 ACRO199340025
Lead(II) oxide 99.7%
Lead (II) oxide
Formula: PbO
Boiling Pt: 1470 °C
Melting Pt: 886 °C
Density: 9,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 9,III
Merck Index: 13,05431

Order Now

Specification Test Results

Appearance Yellow Powder
Titration Complexometric ≥99.6 %
Loss on drying ≤0.1 % (105°C)
Copper (Cu) ≤10 ppm
Iron (Fe) ≤20 ppm
Lead (Pb) ≤0.01 %
Silicon (Si) ≤20 ppm
Lead oxide (Pb3O4) ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR