Dowex® 1X2 (Cl) 200-400 mesh ion-exchange resin

Supplier: Thermo Fisher Scientific

202971000
ACRO202971000EA 82 EUR
ACRO202971000
Dowex® 1X2 (Cl) 200-400 mesh ion-exchange resin
Dowex® 1X2 (Cl)

CAS No.: 9085-42-1

Storage Temperature: Ambient
MDL Number: MFCD00132715
CAS Number: 9085-42-1

Order Now

Specification Test Results

Appearance Beads
Particle size ≤15 % (> 210 µm)
Wet volume capacity ≥0.6 meq/ml (Cl)
Water retention capacity 65 to 75 % (Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR