1-Pentanesulphonic acid sodium salt 98+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate

206110050 206110250 206111000
ACRO206110050EA 39.9 EUR
ACRO206110050 ACRO206110250 ACRO206111000
1-Pentanesulphonic acid sodium salt 98+%
1-Pentanesulphonic acid sodium salt
Formula: C₅H₁₁O₃SNa
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (1 g, 120°C)
Solubility (10 % in water at 25°C) Clear colorless
Titration after Ion exchange ≥98.0 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR