Sodium peroxide 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: di-Sodium peroxide

207700250 207701000 207705000 207700025
ACRO207700250EA 39.8 EUR
ACRO207700250 ACRO207701000 ACRO207705000 ACRO207700025
Sodium peroxide 96%
Sodium peroxide
Formula: Na₂O₂
Melting Pt: 460 °C (dec.)
Density: 2.8 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Merck Index: 13,08728

Order Now

Specification Test Results

Appearance Yellow Granules (May contain a small quantity of discolored material from the manufacturing process)
Titration with KMnO4 ≥95 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR