Mercury(I) nitrate dihydrate 98% for analysis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Dimercury dinitrate dihydrate, Mercurous nitrate dihydrate

213130500 213132500 213130010
ACRO213130500EA 107 EUR
ACRO213130500 ACRO213132500 ACRO213130010
Mercury(I) nitrate dihydrate 98% for analysis
Mercury (I) nitrate dihydrate
Formula: Hg₂N₂O₆·2H₂O
MW: 561.22 g/mol
Melting Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00150812
CAS Number: 7782-86-7
EINECS: 233-886-4
UN: 1627
ADR: 6.1 II
Merck Index: 13,05923

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Assay ≥97.5 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Mercuric Mercury (as Hg) ≤0.5 %
Insoluble matter ≤0.005 % (in dilute HNO3)
Residue after reduction ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR