PIPES disodium salt 99%

Supplier: Thermo Fisher Scientific

Synonyms: Piperazine-N,N'-bis-2-ethanesulphonic acid disodium salt, Piperazine-1,4-bis(2-ethanesulphonic acid) disodium salt, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid disodium salt, PIPES-Na2, 1,4-Piperazinediethanesulphonic acid disodium salt, Disodium piperazine-1,4-diethanesulphonate

215090250 215091000 215092500
ACRO215090250EA 61 EUR
ACRO215090250 ACRO215091000 ACRO215092500
PIPES disodium salt 99%
PIPES disodium salt
Formula: C₈H₁₆N₂Na₂O₆S₂
Storage Temperature: Ambient
MDL Number: MFCD00064349
CAS Number: 76836-02-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤5 %
Solubility (20 % in water) Clear colorless
UV at 280 nm A: ≤0.050
pH 9.2 to 10.0 (1 % in water at 25°C)
Titration with HCl 98.5 to 101.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR