Calcium sulphate, Drierite™ 6 mesh, without indicator

Supplier: Thermo Fisher Scientific

219040020 219045000 219040100
ACRO219040020EA 149 EUR
ACRO219040020 ACRO219045000 ACRO219040100
Calcium sulphate, Drierite™ 6 mesh, without indicator
Calcium sulphate
Formula: CaO₄S
Melting Pt: 1450 °C
Density: 2,964 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Appearance White to greyish-white Irregular granules
Water ≤0.3 %
Particle size 6 mesh
Composition ≥99.5 % (CaSO4) Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR