Zinc, granular 30 mesh

Supplier: Thermo Fisher Scientific
Danger

222615000 222610025 222611000
ACRO222615000EA 110 EUR
ACRO222615000 ACRO222610025 ACRO222611000
Zinc, granular 30 mesh
Zinc
Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7,14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
Merck Index: 12,10255

Order Now

Specification Test Results

Appearance Silver grey Granules or filings
Titration Complexometric ≥99.8 %
Particle size 30 mesh
Suitability (For determination of As) Passes test
Iron (Fe) ≤0.01 %
Lead (Pb) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR