L-Histidinol dihydrochloride 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: L-Histidinol·2 HCl, (S)-2-Amino-3-(1H-imidazol-4-yl)propan-1-ol dihydrochloride

228830010 228830050
ACRO228830010EA 112 EUR
ACRO228830010 ACRO228830050
L-Histidinol dihydrochloride 99%
L-Histidinol dihydrochloride

CAS No.: 1596-64-1

Formula: C₆H₁₃Cl₂N₃O
Melting Pt: 193…199 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00078058
CAS Number: 1596-64-1

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 194 to 200 °C
Titration Argentometric ≥98.5 %
Specific optical rotation -2.0° to -3.3° (20 °C, 589 nm) (c=2, water)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR