Agarose, powder, pure for biochemistry (Mr = - 0.17)

Supplier: Thermo Fisher Scientific

239020250 239021000 239025000
ACRO239020250EA 80.6 EUR
ACRO239020250 ACRO239021000 ACRO239025000
Agarose, powder, pure for biochemistry (Mr = - 0.17)
Agarose
Melting Pt: 60 - 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤7 % (5 g, 110°C, 24 h)
Gel strength ≥2000 g/cm² (1.5 % gel)
Gel point 34.5 to 37.5°C (1.5 % gel)
Electroendosmosis 0.16 to 0.19 (-Mr)
Ash ≤0.5 %
Sulfate (SO4) ≤0.25 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR