2,3-Dichloro-5-(trifluoromethyl)pyridine 97%

Supplier: Thermo Fisher Scientific
Danger

241810250 241811000
ACRO241810250EA 35.3 EUR
ACRO241810250 ACRO241811000
2,3-Dichloro-5-(trifluoromethyl)pyridine 97%
2,3-Dichloro-5-(trifluoromethyl)pyridine
Formula: C₆H₂Cl₂F₃N
Boiling Pt: 176 °C (760 mmHg)
Melting Pt: 8…9 °C
Density: 1.549 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00042243
CAS Number: 69045-84-7
EINECS: 410-340-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4745 to 1.4765 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR