Ammonia solution 25% in water, extra pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

255210025 255210010 255210100
ACRO255210025EA 73.5 EUR
ACRO255210025 ACRO255210010 ACRO255210100
Ammonia solution 25% in water, extra pure
Ammonia solution
Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –57 °C
Density: 0.91 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011418
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl ≥25 % (As NH3)
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR