Dimcarb (dimethylammonium dimethylcarbamate) (Carbon dioxide:Dimethylamine) <2:1

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-Methylmethanamine dimethylcarbamate, Dimethylamine carbon dioxide complex

292901000 292905000 292900250
ACRO292901000EA 66.2 EUR
ACRO292901000 ACRO292905000 ACRO292900250
Dimcarb (dimethylammonium dimethylcarbamate) (Carbon dioxide:Dimethylamine) <2:1
Dimcarb (Dimethylammonium dimethylcarbamate)
Formula: (CH₃)₂NH · (CH₃)₂NCOOH
Boiling Pt: 60…61 °C (760 mmHg)
Density: 1.05 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00029888
CAS Number: 4137-10-4
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4560 (20°C, 589 nm)
Water ≤1 %
Ratio < 2.0/1 Carbon dioxide/Dimethylamine


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR