Sebacoyl dichloride 92%, Technical Grade

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Sebacoyl chloride

294902500 294905000 294900500
ACRO294902500EA 180 EUR
ACRO294902500 ACRO294905000 ACRO294900500
Sebacoyl dichloride 92%, Technical Grade
Sebacoyl dichloride
Formula: C₁₀H₁₆Cl₂O₂
Boiling Pt: 220 °C (75 mmHg)
Melting Pt: –2.5 °C
Density: 1.121 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000770
CAS Number: 111-19-3
EINECS: 203-843-4
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow to golden to brown Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4700 (20°C, 589 nm)
GC ≥90 %
Titration RCOCl ≥90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR