4-(Methylthio)aniline 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-(Methylmercapto)aniline

309830050
ACRO309830050EA 62.3 EUR
ACRO309830050
4-(Methylthio)aniline 98%
4-(Methylthio)aniline
Formula: C₇H₉NS
Boiling Pt: 272…273 °C
Melting Pt: 156…158 °C
Density: 1.119 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00007889
CAS Number: 104-96-1
EINECS: 203-256-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear light yellow to orange to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.6380 to 1.6420 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR