1,2-Bis(chlorodimethylsilyl)ethane 96%

Supplier: Thermo Fisher Scientific
Danger

313260050 313260250
ACRO313260050EA 24.3 EUR
ACRO313260050 ACRO313260250
1,2-Bis(chlorodimethylsilyl)ethane 96%
1,2-Bis(chlorodimethylsilyl)ethane

Appearance: Transparent Low melting solid

Formula: C₆H₁₆Cl₂Si₂
Boiling Pt: 198…200 °C (760 mmHg)
Melting Pt: 35…38 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000505
CAS Number: 13528-93-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Transparent Low melting solid
Infrared spectrum Conforms
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR