Bismuth(III) nitrate pentahydrate 99.999% (trace metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Bismuth trinitrate pentahydrate

315150250 315150050 315151000
ACRO315150250EA 109 EUR
ACRO315150250 ACRO315150050 ACRO315151000
Bismuth(III) nitrate pentahydrate 99.999% (trace metals basis)
Bismuth (III) nitrate pentahydrate
Formula: Bi(NO₃)₃·5H₂O
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: 30 °C
Density: 2,736 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149157
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystals
Total trace metal impurities ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR