Chromium(II) chloride 99.9% (trace metals basis)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Chromium dichloride

315190050 315190010 315190250
ACRO315190050EA 204 EUR
ACRO315190050 ACRO315190010 ACRO315190250
Chromium(II) chloride 99.9% (trace metals basis)
Chromium (II) chloride
Formula: Cl₂Cr
Boiling Pt: 1302 °C (1013 hPa)
Melting Pt: 824 °C
Density: 2,87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010947
CAS Number: 10049-05-5
EINECS: 233-163-3
ADR: 8,III
Merck Index: 13,02264

Order Now

Specification Test Results

Appearance Off-white to grey-green Powder
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR