n-Octane 97%, pure

Supplier: Thermo Fisher Scientific
Danger

325950025 325950010 325950250
ACRO325950025EA 332 EUR
ACRO325950025 ACRO325950010 ACRO325950250
n-Octane 97%, pure
n-Octane
Formula: H₃C(CH₂)₆CH₃
Boiling Pt: 125…127 °C (760 mmHg)
Melting Pt: –57 °C
Density: 0.708 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009556
CAS Number: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,II
Merck Index: 13,06782
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68adf890d68728825d4a2fa4b032700b93f710b8&vid=b00a2925902bc425c80f8ebb3f0e7b456c800ae3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3965 to 1.3985 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR