2-Propanol 99.5% Molecular_Biology DNAse_RNAse_PROTEase_free

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

327270010 327272500
ACRO327270010EA 133 EUR
ACRO327270010 ACRO327272500
2-Propanol 99.5% Molecular_Biology DNAse_RNAse_PROTEase_free
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 81…83 °C (760 mmHg)
Melting Pt: –89.5 °C
Density: 0.785 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 % (Coulometric)
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Dnase,Rnase,Protease activity none detected
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Nickel (Ni) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm
Acidity (CH3COOH) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR