4-Amino-6-methoxypyrimidine 97%

Supplier: Thermo Fisher Scientific
Warning

337450050 337450250
ACRO337450050EA 68.6 EUR
ACRO337450050 ACRO337450250
4-Amino-6-methoxypyrimidine 97%
4-Amino-6-methoxypyrimidine

Appearance: White to yellow Crystalline powder

Formula: C₅H₇N₃O
Melting Pt: 154…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00129983
CAS Number: 696-45-7

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 154 to 159 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR