N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine

344300050 344300250
ACRO344300050EA 128 EUR
ACRO344300050 ACRO344300250
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 96%
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine

CAS No.: 93102-05-7

Formula: C₁₃H₂₃NOSi
MW: 237.42 g/mol
Boiling Pt: 76 °C (.3 mmHg)
Density: 0.928 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00674005
CAS Number: 93102-05-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4940 (20°C, 589 nm)
Elemental Analysis ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR