Scandium(III) trifluoromethanesulphonate 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Scandium(III) trifluoromethanesulphonate, Scandium triflate

363890010 363890050
ACRO363890010EA 95.6 EUR
ACRO363890010 ACRO363890050
Scandium(III) trifluoromethanesulphonate 95%
Scandium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Sc
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00192433
CAS Number: 144026-79-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Elemental Analysis ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR