2-Methylpyrrolidine 98%

Supplier: Thermo Fisher Scientific
Danger

365390020 365390100
ACRO365390020EA 114 EUR
ACRO365390020 ACRO365390100
2-Methylpyrrolidine 98%
2-Methylpyrrolidine
Formula: C₅H₁₁N
Boiling Pt: 98…100 °C
Density: 0.88 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00014491
CAS Number: 765-38-8
EINECS: 212-144-3
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4370 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR