Methyl-3-(bromomethyl)benzoate 95%

Supplier: Thermo Fisher Scientific
Danger

368630050 368630250 368631000
ACRO368630050EA 72.9 EUR
ACRO368630050 ACRO368630250 ACRO368631000
Methyl-3-(bromomethyl)benzoate 95%
Methyl 3-(bromomethyl)benzoate
Formula: C₉H₉BrO₂
Boiling Pt: 135 °C (2 mmHg)
Melting Pt: 35…37 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051437
CAS Number: 1129-28-8
EINECS: 419-100-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 41 to 47 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR