Ammonium tetrathiomolybdate 99.95% (trace metals basis)

Supplier: Thermo Fisher Scientific
Warning

389530010 389530100 389530500 389532500
ACRO389530010EA 42.7 EUR
ACRO389530010 ACRO389530100 ACRO389530500 ACRO389532500
Ammonium tetrathiomolybdate 99.95% (trace metals basis)
Ammonium tetrathiomolybdate
Formula: H₁₀MoN₂S₄
Melting Pt: 300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00136013
CAS Number: 15060-55-6

Order Now

Specification Test Results

Appearance Brown to black Powder or crystals
Total trace metal impurities ≤500 ppm
Molybdenum (Mo) 35.5 to 38.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR