N-Boc-ethylenediamine 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-(tert-Butoxycarbonyl)-1,2-diaminoethane, N-(tert-Butoxycarbonyl)-1,2-ethylenediamine, tert-Butyl (2-aminoethyl)carbamate, t-boc-N-EDA, tert-Butyl N-(2-Aminoethyl)carbamate, N-BOC-1,2-diaminoethane

389900010 389900050 389900250
ACRO389900010EA 57 EUR
ACRO389900010 ACRO389900050 ACRO389900250
N-Boc-ethylenediamine 98%
N-Boc-ethylenediamine
Formula: C₇H₁₆N₂O₂
Boiling Pt: 80…84 °C (.3 mmHg)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191871
CAS Number: 57260-73-8
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR