Sodium polyphosphate (Graham's salt), pure

Supplier: Thermo Fisher Scientific
Warning

390930010 390930050 390932500
ACRO390930010EA 55 EUR
ACRO390930010 ACRO390930050 ACRO390932500
Sodium polyphosphate (Graham's salt), pure
Sodium polyphosphate (Graham's salt)
Formula: (NaO₃P)n
Melting Pt: 600 °C
Density: 2,4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134118
CAS Number: 50813-16-6
EINECS: 256-779-4
Merck Index: 15, 8773

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration excess HCl 65 to 70 %
Chloride (Cl) ≤0.03 %
Sulfate (SO4) ≤0.1 %
Iron (Fe) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR