Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

Supplier: Thermo Fisher Scientific
Warning

392140010 392140050
ACRO392140010EA 129 EUR
ACRO392140010 ACRO392140050
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Formula: C₅₅H₄₅ClOP₃Ru
Melting Pt: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00049804
CAS Number: 16971-33-8

Order Now

Specification Test Results

Appearance Cream to tan to brown Powder or crystals
Infrared spectrum Conforms
Ruthenium (Ru) ≥10.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR