Geraniol 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

410900250 410901000 410900050 410905000 410900025
ACRO410900250EA 48.8 EUR
ACRO410900250 ACRO410901000 ACRO410900050 ACRO410905000 ACRO410900025
Geraniol 99%
Geraniol
Formula: C₁₀H₁₈O
Boiling Pt: 229…230 °C (757 mmHg)
Melting Pt: –15 °C
Density: 0.878 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4780 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR