D-(-)-Leucinol 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: D-Leucinol, (R)-(-)-Leucinol, (R)-(-)-2-Amino-4-methylpentan-1-ol, (R)-2-Amino-4-methylpentan-1-ol

413120010 413120050
ACRO413120010EA 38.6 EUR
ACRO413120010 ACRO413120050
D-(-)-Leucinol 97%
D(-)-Leucinol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 199 °C (760 mmHg)
Density: 0.907 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004734
CAS Number: 53448-09-2

Order Now

Specification Test Results

Appearance Clear light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4530 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation -3° to -5° (20°C, 589 nm) (c=9, ethanol)
Titration with HClO4 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR