4-(tert-Butoxycarbonylamino)-1-butanol 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: tert-Butyl (4-hydroxybutyl)carbamate, N-Boc-4-aminobutanol, 4-(Boc-amino)-1-butanol, N-(4-Hydroxybutyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(4-Hydroxybutyl)carbamate

434210010 434210050
ACRO434210010EA 71.8 EUR
ACRO434210010 ACRO434210050
4-(tert-Butoxycarbonylamino)-1-butanol 98%
4-(tert-Butoxycarbonylamino)-1-butanol
Formula: C₉H₁₉NO₃
Melting Pt: 37…40 °C
Density: 1.02 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD01861334
CAS Number: 75178-87-9

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR