3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride

440520050 440520250 440521000
ACRO440520050EA 23.7 EUR
ACRO440520050 ACRO440520250 ACRO440521000
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
Formula: C₁₃H₁₆ClNOS
Melting Pt: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00011959
CAS Number: 4568-71-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 141 to 147 °C
Elemental Analysis 5.06 to 5.32 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR