Nitromethane 96% for HPLC

Supplier: Thermo Fisher Scientific
Warning

445670010 445670025 445671000
ACRO445670010EA 123 EUR
ACRO445670010 ACRO445670025 ACRO445671000
Nitromethane 96% for HPLC
Nitromethane
Formula: CH₃NO₂
Boiling Pt: 101.2 °C (1013 hPa)
Melting Pt: –29 °C
Density: 1.12 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00007400
CAS Number: 75-52-5
UN: 1261
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.03 %
GC ≥95 %
UV at 430 nm A: ≤0.01
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR