Sodium dihydrogen citrate, anhydrous 99% for analysis

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Sodium 3,4-dicarboxy-3-hydroxybutanoate, Sodium citrate monobasic, mono-Sodium citrate, Citric acid monosodium salt, Monosodium Citrate

447692500 447690010
ACRO447692500EA 32.3 EUR
ACRO447692500 ACRO447690010
Sodium dihydrogen citrate, anhydrous 99% for analysis
Sodium dihydrogen citrate
Formula: C₆H₇NaO₇
Melting Pt: 209…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00013067
CAS Number: 18996-35-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying ≤1 %
Solubility Clear colorless
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.005 %
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR