Chloroacetic acid 99+% ACS

Supplier: Thermo Fisher Scientific
Danger

448110025 448111000 448115000
ACRO448110025EA 201 EUR
ACRO448110025 ACRO448111000 ACRO448115000
Chloroacetic acid 99+% ACS
Chloroacetic acid
Formula: ClCH₂COOH
Boiling Pt: 189 °C (760 mmHg)
Melting Pt: 61…63 °C
Density: 1.58 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002683
CAS Number: 79-11-8
UN: 1751
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2516e0c26c6c26f4f6f3656bc5b4256e7cf08207&vid=9c9b7ad094469d9089eb2f0ce88275d1423c2b46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to white Deliquescent crystals or flakes
Sulfated ash ≤0.02 %
Titration with NaOH ≥99.0 %
Substance darkened by H2SO4 passes test
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.02 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.01 %
Carbonyl compounds ≤0.01 % (other)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR