Toluene, technical

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

451970050
ACRO451970050EA 98.1 EUR
ACRO451970050
Toluene, technical
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: –95 °C
Density: 0.866 g/cm³
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
UN: 1294
ADR: 3 II
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Refractive index 1.4900 to 1.5030 (20°C, 589 nm)
Density 0.865 to 0.872 kg/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR