Millon's reagent for the detection of tyrosine

Supplier: Thermo Fisher Scientific
Danger

458541000 458545000
ACRO458541000EA 53.4 EUR
ACRO458541000 ACRO458545000
Millon's reagent for the detection of tyrosine
Millon's reagent
Density: 1.4 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00147623
CAS Number: 10045-94-0
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Density 1.2 to 1.5 kg/l
Sensitivity passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR