Meloxicam 99-101%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Hydroxy-2-methyl-N-(5-methylthiazol-2-yl)-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide

459550050 459550250
ACRO459550050EA 37.2 EUR
ACRO459550050 ACRO459550250
Meloxicam 99-101%
Meloxicam
Formula: C₁₄H₁₃N₃O₄S₂
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00868752
CAS Number: 71125-38-7

Order Now

Specification Test Results

Appearance Light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 4 hr)
Residue after ignition ≤0.1 %
Titration with HClO4 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR