Chalcogran mixture of isomers 96%

Supplier: Thermo Fisher Scientific

Synonyms: 2-Ethyl-1,6-dioxaspiro[4,4]-nonane

461410010
ACRO461410010EA 241 EUR
ACRO461410010
Chalcogran mixture of isomers 96%
Chalcogran
Formula: C₉H₁₆O₂
Boiling Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01861911
CAS Number: 38401-84-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4465 (20°C, 589 nm)
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR